Water with 0,1% (v/v) formic acid, BAKER ANALYZED® for LC-MS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Management of Change (MOC) category = NI
BAKR9834-02
BAKR9834-03
CAJT9834-2CS
896.68
CAD
CAJT9834-2
CAJT9834-3
Water with 0,1% (v/v) formic acid, BAKER ANALYZED® for LC-MS, J.T.Baker®
Water with 0,1% (v/v) formic acid
Clear, colorless liquid. For use in liquid chromatography and mass spectrometry.
Storage Temperature:
Ambient |
EINECS:
200-579-1 REACH: 01-2119491174-37-XXXX |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Appearance | Passes Test |
Formic Acid | 0.095 - 0.105 % |
Gradient Elution Test (a.u.) - 254 nm | ≤0.01 |
Residue after Evaporation | ≤1.0 ppm |
LC/MS Suitability - Largest Response on ESI- Positive Mode (as Reserpine) | ≤50 ppb |
Trace Impurities - Aluminum (Al) | ≤50.0 ppb |
Trace Impurities - * Calcium (Ca) | ≤50.0 ppb |
Trace Impurities - Iron (Fe) | ≤50.0 ppb |
Trace Impurities - Magnesium (Mg) | ≤50.0 ppb |
Trace Impurities - Potassium (K) | ≤50 ppb |
Trace Impurities - * Sodium (Na) | ≤50.0 ppb |
* May change over time due to extraction from glass container. | |
Filtered through a 0.2 micron filter. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...