Formamide ≥99.5% (by GC), Ultrapure Bioreagent for biochemistry, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Carbamaldehyde

Management of Change (MOC) category = N

BAKR4028-00 BAKR4028-01
CAJT4028-0EA 188.75 CAD
CAJT4028-0 CAJT4028-1
Formamide ≥99.5% (by GC), Ultrapure Bioreagent for biochemistry, J.T.Baker®
Formamide

For denaturing nucleic acids prior to analysis.


ULTRAPURE BIOREAGENT—A line of high purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography. Where applicable, tests include DNase, RNase, Protease, heavy metals and insoluble matter.

Formula: HCONH₂
MW: 45.04 g/mol
Boiling Pt: 210
Melting Pt: 2
Density: 1.1334 g/cm³ (20 °C)
Flash Pt: 79
Storage Temperature: Refrigerator
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (HCONH₂) (by GC) ≥99.5 %
Absorbance of a 1M Solution - 280 nm ≤0.100
Absorbance of a 1M Solution - Color (APHA) ≤10
Absorbance of a 1M Solution - Conductivity, mS/cm (1:10), For Information Only
Freezing Point 2.0 - 3.0 °C
Trace Impurities - Copper (Cu) ≤0.1 ppm
Trace Impurities - Iron (Fe) ≤0.5 ppm
Trace Impurities - Lead (Pb) ≤0.5 ppm
Trace Impurities - Zinc (Zn) ≤0.5 ppm
Packaged under Nitrogen
Deionization of material is not required.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR