Urea ≥99.5%, OmniPur®, Millipore®

Supplier: MilliporeSigma

9510-500GM 9530-5KG 9550-25KG
CA-EM9510EA 164.18 CAD
CA-EM9510 CA-EM9530 CAEM9550
Urea ≥99.5%, OmniPur®, Millipore®
Urea

Ideal for protein unfolding/denaturation.

Formula: NH₂CONH₂
Boiling Pt: 197 °C (1013 hPa)
Melting Pt: 132…135 °C (lit.)
Density: 1.335 g/ml (25°C) (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00008022
CAS Number: 57-13-6
EINECS: 200-315-5
REACH: 01-2119463277-33
Merck Index: 13,09935
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a9682f71b212a449398841e2beace2d7500b4710&vid=d6057a7ec3d606031d338a300b5d5db30b52d861&prid=web&clid=SAPDAM

Order Now

Specification Test Results

A (260 nm, 0.1 M, water) 0.01AU max
A (280 nm, 0.1 M, water) 0.01AU max
Appearance White crystalline powder
Assay 99.5% min
DNase None detected
Heavy metals (as Pb) 0.0010% max
Phosphatase None detected
Protease None detected
RNase None detected


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR