Florisil®, BAKER ANALYZED® (60-100 Mesh), activated, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Talc, Magnesium silicate, activated, Silicic acid magnesium salt, Florisil

M.O.C. Category N

BAKR3372-07 BAKR3372-08
CAJT3372-7EA 753.71 CAD
CAJT3372-7 CAJT3372-8
Florisil®, BAKER ANALYZED® (60-100 Mesh), activated, J.T.Baker®
Magnesium silicate

For chromatographic cleanup of pesticide residue.


  • Activated at 675 °C

Formula: MgO₃Si
MW: 100.38 g/mol
Density: 2.51 Kg/L
MDL Number: MFCD00078249
CAS Number: 1343-88-0
EINECS: 215-681-1

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Mesh - On U.S. No. 60 Sieve ≤10 %
Mesh - Thru U.S. No. 100 Sieve ≤10 %
Product may contain black particles that are produced during the manufacturing process.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR