Formaldehyde ≥37% in aqueous solution stabilised BP, Ph. Eur., USP analytical reagent, SAFC®

Supplier: EMD MILLIPORE – PCS CA EXCEP
Danger

Synonyms: Paraformaldehyde aqueous solution, Methanal solution, Methylaldehyde solution, Formol, Formaline solution, Formalin, Methylaldehyde

Stabilization: Stabilised with methanol 10%
M-Clarity™ quality level = MQ400

1.04002.2500 1.04002.1000 1.04002.9025 1.04002.9200
CA1.04002.2500EA 3248.45 CAD
CA1.04002.2500 CA1.04002.1000 CA1.04002.9025 CA1.04002.9200
Formaldehyde ≥37% in aqueous solution stabilised BP, Ph. Eur., USP analytical reagent, SAFC®
Formaldehyde

EMSURE® (formerly GR) is a premium grade of reagents for wet chemical analysis, adhering to ACS, ISO, and/or Reag. Ph.Eur quality specifications.


In North America 1.04002.2500 replaces and is identical to 1.04002.2507 In North America 1.04002.9200 replaces and is identical to 1.04002.9187

Formula: CH₂O
Boiling Pt: 93…96 °C (1013 hPa)
Melting Pt: <–15 °C
Density: 1.09 g/cm3 (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
EINECS: 200-001-8
UN: 1198
ADR: 8,III

Order Now

Specification Test Results

Assay (iodometric) 37.0 - 38.0 %
Identity conforms
Appearance of solution Passes test
Acidity (Ph Eur, BP) Passes test
Acidity (according to USP) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR