Formic acid 0,1% v/v acetonitrile HPLC_mobile_phase, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Aminic Acid, Methanoic acid

FX0437-NP20 FX0437P-1
CAFX0437-NP20EA 4141 CAD
CAFX0437-NP20 CAFX0437P-1
Formic acid 0,1% v/v acetonitrile HPLC_mobile_phase, Supelco®
Formic acid
Formula: HCO₂H
MW: 46.03
Density: 1.22
Flash Pt: 48
Storage Temperature: Ambient
MDL Number: MFCD00003297
CAS Number: 64-18-6
EINECS: 200-579-1
UN: 1648
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a6cee80f8dc5a4afaf0be47215f1cf258eef7cfd&vid=a77e0bed84455e37172a184511237ad1d178fd11&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (HCOOH) 0.095-0.105% (v/v)
Color (APHA) 10 max
Density at 25°C Determine
Filtered through 0.2 µm filter To pass test
Form Clear liquid
Gradient at 280 nm 0.005AU max
Residue after evaporation 5ppm max
UV Abs. at 230 nm 1.00AU max
UV Abs. at 240 nm 0.50AU max
UV Abs. at 250 nm 0.05AU max
UV Abs. at 280 nm and higher 0.05AU max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR