Formamide ≥99.5%, GR ACS, Supelco®

Supplier: MilliporeSigma

Synonyms: Carbamaldehyde

FX0420-6 FX0420-6
CA-FX0420-6CS 6382.44 CAD
EM-FX0420-6 CA-FX0420-6
Formamide ≥99.5%, GR ACS, Supelco®
Formamide

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: HCONH₂
MW: 45.04 g/mol
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1.1334 g/cm³ (20 °C)
Flash Pt: 79 °C
MDL Number: MFCD00007941
CAS Number: 75-12-7
Merck Index: 13,04261
https://ca.vwr.com/stibo/web/std.lang.all/25/02/2302502.jpg

Order Now

Specification Test Results

Assay (GC) 99.5% min
Appearance Colorless, slightly viscous liquid
Color (APHA) 10 max
Freezing point 2.0-3.0°C
Identity (IR-spectrum) Conforms
Water (H2O) 0.25% max

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR