Formamide ≥99.0%, OmniPur®, deionised, Millipore®

Supplier: MilliporeSigma
Danger

Synonyms: Carbamaldehyde

4610-100ML 4650-500ML 4670-4L
CA-EM4610EA 131.4 CAD
CA-EM4610 CA-EM4650 CAEM4670
Formamide ≥99.0%, OmniPur®, deionised, Millipore®
Formamide

For molecular biology. DNase-, RNase-, protease-, and phosphatase-free.

Formula: HCONH₂
Boiling Pt: 210 °C (1013 hPa)
Melting Pt: 2 °C
Density: 1.1334 g/cm³ (20 °C)
Flash Pt: 79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007941
CAS Number: 75-12-7
EINECS: 200-842-0
Merck Index: 13,04261
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f381691a96baf5054ca8291e1fa9e9603b687e31&vid=e54dda1a985d7a4f9485c2cc5233498087e43f88&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR