Formic Acid/Acetonitrile Blend, LabReady™ LC-MS, Solvent Blend LC-MS, HPLC, Burdick & Jackson™
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
LC441-1
LC441-2.5
CABJLC441-1CS
1403.18
CAD
CABJLC441-1
CABJLC441-2.5
Formic Acid/Acetonitrile Blend, LabReady™ LC-MS, Solvent Blend LC-MS, HPLC, Burdick & Jackson™
Acetonitrile with 0,1% (v/v) Formic acid
LabReady™ Solvent Blends
- Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
- Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
- Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
- Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk
Boiling Pt:
82 °C (1.013 hPa) Melting Pt: –46 °C Density: 0.7767 g/cm³ (25 °C) Flash Pt: 8 °C Storage Temperature: Ambient |
EINECS:
200-579-1 UN: 1648 ADR: 3 II |