Water with 0,1% (v/v) formic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Supplier: HONEYWELL BURDICK & JACKSON CA
NP452-20
452-4
NS452-20
452-4
NP452-20
CABJNP452-20EA
16255.25
CAD
CABJNP452-20
BJNS452-20
BJ452-4
BJNP452-20
CABJ452-4
Water with 0,1% (v/v) formic acid, LabReady™, Solvent Blend for HPLC, Burdick & Jackson™
Water with 0,1% (v/v) formic acid
LabReady™ Solvent Blends
- Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
- Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
- Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
- Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk
Specification Test Results
Formic Acid (v/v) | 0.09 - 0.11% |
UV Cutoff | <215 nm |
Residue | < 1 mg/L |
Color (APHA) | < 10 |
HPLC Gradient @ 215 nm | < 0.020 AU |
HPLC Gradient @ 254 nm | < 0.010 AU |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...