, LabReady™ HPLC, Solvent Blend HPLC, Burdick & Jackson™

Supplier: HONEYWELL BURDICK & JACKSON CA
Danger

NS441-20 441-4
CABJNP441-20EA 30272.82 CAD
CABJNP441-20 CABJNS441-20 CABJ441-4
, LabReady™ HPLC, Solvent Blend HPLC, Burdick & Jackson™
Acetonitrile with 0,1% (v/v) Formic acid

LabReady™ Solvent Blends


  • Using custom blends not only helps you reduce waste through human error, but also cuts disposal costs for hazardous materials used during blend preparation
  • Honeywell Research Chemicals uses proprietary, closed-loop blending technology, which eliminates variation, ensuring your blends are as precise as possible
  • Honeywell LabReady Blends undergo full safety analyses, with a safety data sheet provided for every blended product
  • Honeywell’s high-precision blending technology reduces contamination and exposure during the blending process, helping to minimize environmental risk

Boiling Pt: 82 °C (1.013 hPa)
Melting Pt: –46 °C
Density: 0.7822 g/cm³ (20 °C)
Flash Pt: 8 °C
Storage Temperature: Ambient
EINECS: 200-579-1
UN: 1648
ADR: 3 II

Order Now

Specification Test Results

Formic Acid (v/v) 0.09 - 0.11%
UV Cutoff <224 nm
Residue < 1 mg/L
Color (APHA) < 10
HPLC Gradient @ 215 nm < 0.020 AU
HPLC Gradient @ 254 nm < 0.010 AU


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR