Formalin 10% neutral buffered, HiPur™
Supplier: RICHARD-ALLAN SCIENTIFIC CA
|
Danger
|
Synonyms:
Paraformaldehyde aqueous solution, Formaldehyde, Methylaldehyde, Formol
9400-1
9400-5
CA48218-698CS
324.69
CAD
CA48218-698
CA48218-700
Formalin 10% neutral buffered, HiPur™
Formalin
Choose the option that best fits your workflow, HiPur™ 10% neutral buffered Formalin is available in one and five gallon sizes.
Contains neutral phosphate buffer and methanol.
Formula:
CH₂O MW: 30.03 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00003274 CAS Number: 50-00-0 EINECS: 200-001-8 UN: 3334 ADR: 9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...