(4-Aminophenyl)acetic acid ethyl ester ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 4-aminophenylacetate, Ethyl (4-aminophenyl)acetate

A1289-25G A1289-5G
TCA1289-025GEA 1161.3 CAD
TCA1289-025G TCA1289-5G
(4-Aminophenyl)acetic acid ethyl ester ≥98.0%
(4-Aminophenyl)acetic acid ethyl ester

CAS Number: 5438-70-0
MDL Number: MFCD00017569
Molecular Formula: C10H13NO2
Molecular Weight: 179.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 50


Other Product Names / Synonyms: 4-Aminophenylacetic Acid Ethyl Ester

Formula: C₁₀H₁₃NO₂
MW: 179.22 g/mol
Melting Pt: 50 °C
MDL Number: MFCD00017569
CAS Number: 5438-70-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3c3e90e9772965c3458a484ee9e59d3a25a58f03&vid=3c3e90e9772965c3458a484ee9e59d3a25a58f03&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR