Eriochrome Black T 0,5% (w/v) in 2-methoxyethanol for hardness of water determinations

Supplier: Ricca Chemical

Synonyms: 1-Naphthalenesulfonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1), 1-Naphthalenesulphonic acid, 3-hydroxy-4-[2-(1-hydroxy-2-naphthalenyl)diazenyl]-7-nitro-, sodium salt (1:1), Eriochrome black T, Indicator, EBT, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulphonate, Solochrome Black T, Sodium 3-hydroxy-4-(2-hydroxy-1-naphthylazo)-7-nitro-1-naphthalenesulfonate

R9300000-500C R9300000-120C
RC930016EA 129.56 CAD
RC930016 RC93004
Eriochrome Black T 0,5% (w/v) in 2-methoxyethanol for hardness of water determinations
Eriochrome Black T

In 2-Methoxyethanol.

Formula: C₂₀H₁₂N₃NaO₇S
MW: 461.39 g/mol
MDL Number: MFCD00003935
CAS Number: 1787-61-7
UN: 3077
ADR: 9 III
Merck Index: 13,03702

Order Now

Specification Test Results

Appearance Dark red, Methoxyethanol odor
Suitability for Use Wine Red (excess Ca or Mg) - Pure Blue (excess EDTA)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR