Reagent alcohol, anhydrous ACS (denatured with 5% isopropanol and 5% methanol)

Supplier: Ricca Chemical
Danger

R6590000-10F R6590000-3.8A
RCR6590000-10FEA 654.12 CAD
RCR6590000-10F RCR6590000-4A RCR65900003.8A
Reagent alcohol, anhydrous ACS (denatured with 5% isopropanol and 5% methanol)
Alcohol

Denatured with 4 to 6% methanol and 4 to 6% 2-propanol.


Ordering information: The purchase of Reagent Alcohol in containers larger than 4 liters requires a Letterhead Permit approval by the United States Treasury Tax and Trade Bureau (TTB).

Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear, colorless, alcohol odor
Color max. 10
Ethyl Alcohol 90.5 ± 1.0 % (v/v)
Isopropyl Alcohol 5.0 ± 1.0 % (v/v)
Methyl Alcohol 4.5 ± 0.5 % (v/v)
Residue on Evaporation max. 0.001 %
Water max. 0.5 % (w/w)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR