EDTA disodium salt 0.01 M (M/100), StandARd® Volumetric, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

M.O.C. Category NI

MACRH105-05 MACRH105-07
CAMKH10505EA 83.17 CAD
CAMKH10505 CAMKH10507
EDTA disodium salt 0.01 M (M/100), StandARd® Volumetric, Macron Fine Chemicals™
EDTA disodium salt

For calcium hardness determination.

Formula: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS Number: 139-33-3
EINECS: 205-358-3
Merck Index: 12,03556

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Molarity 0.0095 - 0.0105


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR