EDTA disodium salt 0.1 M (M/10), StandARd® Volumetric, Macron Fine Chemicals™

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Idranal III, Edetic acid disodium salt, Kestranal® 2S, Ethylenedinitrilotetra-acetic acid disodium salt, Disodium dihydrogen ethylenediaminetetraacetate, Ethylenediaminetetra-acetic acid disodium salt

M.O.C. Category N

H100-05 H100-07
MKH10005CS 657.33 CAD
MKH10005 MKH10007 CAMKH10005
EDTA disodium salt 0.1 M (M/10), StandARd® Volumetric, Macron Fine Chemicals™
EDTA disodium salt
Formula: C₁₀H₁₄N₂Na₂O₈
MW: 336.21 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00070672
CAS Number: 139-33-3
EINECS: 205-358-3
Merck Index: 12,03556

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Molarity 0.095 - 0.105


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR