Reagent alcohol absolute 94,0-96,0, AR® ACS (IPA 5% + MeoH 5%), Macron Fine Chemicals™

Supplier: VWR International
Danger

Synonyms: Reagent alcohol

M.O.C. Category N

CAMK701902 MACR7019-10 MACR7019-28
CAMK701902CS 1434.53 CAD
CAMK701902 CAMK701910 CAMK7019-28
Reagent alcohol absolute 94,0-96,0, AR® ACS (IPA 5% + MeoH 5%), Macron Fine Chemicals™
Alcohol

GMP manufactured.


  • Total alcohol: 99.5%
  • Ethyl alcohol: 90%
  • Denaturant: 3.5–5.5% methanol and 4.0–6.0% 2-propanol


Ordering information: The purchase of Reagent Alcohol in containers larger than 4 liters requires a Letterhead Permit approval by the United States Treasury Tax and Trade Bureau (TTB).

Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1987
ADR: 3 II

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Specially Denatured Alcohol Formula 3A (200 proof)(v/v) 94.0 - 96.0 %
Ethanol (v/v) 88.5 - 92.5 %
Methanol (v/v) 3.5 - 5.5 %
Isopropyl Alcohol (2-propanol)(v/v) 4.0 - 6.0 %
Total Alcohol (v/v) ≥98.0 %
Color (APHA) ≤10
Residue after Evaporation ≤0.001 %
Specific Gravity at 25°C 0.782 - 0.793
Water (H₂O) ≤0.5 %
SDA Formula 3A consists of 5 volumes of methanol and 100 volumes of 200
proof ethanol. Product Information (not specifications): Appearance
(clear, colorless liquid)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR