Water, Reagent Grade, distilled

Supplier: Ricca Chemical

R9180000-4C R9180000-1C R9180000-20F R9180000-55E R9180000-100C
RC9180-1EA 237 CAD
RC9180-1 RC9180-32 RCR9180000-20F RCR9180000-55E RCR9180000100C
Water, Reagent Grade, distilled
Water

The high purity Water feedstock for this product is manufactured by a continuous process that has been validated to produce high purity Water meeting, as a minimum, ACS Reagent Grade, ASTM Type I, and ASTM Type II specifications. This high purity Water is then further purified by an added distillation step and filled into specially cleaned containers. This product has a low microbial count but is not guaranteed to be sterile. The conductivity specification for this product is equivalent to a minimum resistivity of 0.67 megohm-cm.

Formula: H₂O
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0.997 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Appearance Colorless liquid
Conductivity at 25°C (Method: SQCP031) max 1.5 µS/cm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR