Water ACS, Reagent Grade for HPLC, for UV spectrophotometry

Supplier: Ricca Chemical

Synonyms: Hydrogen oxide

R9153000-1C R9153000-4C
RC9153-32EA 93.22 CAD
RC9153-32 RC9153-1
Water ACS, Reagent Grade for HPLC, for UV spectrophotometry
Water

Suitable for ultraviolet spectrophotometry and liquid chromatography.

Formula: H₂O
Boiling Pt: 100 °C
Melting Pt: 0 °C
Density: 0.997 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Chloride (Cl) max 0.4 ppm
Conductivity at 25°C (Method: SQCP031) max 0.056 µS/cm
Heavy Metals (as Pb) max 0.01 ppm
LC Suitability (Absorbance) To Pass Test
LC Suitability (Gradient Elution Test) To Pass Test
Nitrate (NO₃) max 0.4 ppm
Organic Carbon (TOC) max 50 ppb
Oxidizable Substances (Permanganate Retention) To Pass Test
Phosphate (PO₄) max 1.0 ppm
Silicate (as SiO₂) max 0.01 ppm
Sulfate (SO₄) max 1.0 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR