EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥98%, Laboratory Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Warning
|
Management of Change (MOC) category = N
BAKRL657-05
BAKRL657-06
CAJTL657-5EA
2829.04
CAD
CAJTL657-5
CAJTL657-6
EGTA (ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid) ≥98%, Laboratory Reagent, J.T.Baker®
EGTA (Ethylene glycol bis(2-aminoethyl ether)-N,N,N',N'-tetraacetic acid)
Formula:
C₁₄H₂₄N₂O₁₀ Melting Pt: 240…244 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004291 CAS Number: 67-42-5 EINECS: 200-651-2 Merck Index: 13,03560 |
Specification Test Results
For Laboratory,Research,or Manufacturing Use | |
Assay (C₁₄H₂₄N₂O₁₀) | ≥98 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...