Water, BAKER ANALYZED® HPLC for HPLC, for LC-MS, J.T.Baker®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC

Synonyms: Hydrogen oxide

Management of Change (MOC) category = NI

BAKR4218-02 4218-03
CAJT4218-2CS 655.98 CAD
CAJT4218-2 CAJT4218-3 JT4218-3 CAJT4218-88
Water, BAKER ANALYZED® HPLC for HPLC, for LC-MS, J.T.Baker®
Water

For use in high-performance liquid chromatography. Free from organic compounds. Lot analysis on label.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
UV Absorbance on Gradient Elution - Largest Peak at 254nm (au) ≤0.001
UV Absorbance on Gradient Elution - Largest Peak at 205nm (au), for
Information Only
Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission ≤0.1 ppb
Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities ≤0.2 ppb
Filtration Test Passes Test
pH, For Information Only
Residue after Evaporation ≤2.0 ppm
Filtered through a 0.2 micron filter.
Appearance - A clear, colorless, odorless liquid.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR