Water, OmniPur®, WFI (water for injection) quality cell culture tested, sterile, Millipore®

Supplier: MilliporeSigma

Synonyms: Hydrogen oxide

4.86505.5000 4.86505.9010 4.86505.9020 4.86505.9200
CA4.86505.5000EA 636.46 CAD
CA4.86505.5000 CA4.86505.9010 CA4.86505.9020 CA4.86505.9200 CA4.86505.0500 CA4.86505.1000
Water, OmniPur®, WFI (water for injection) quality cell culture tested, sterile, Millipore®
Water

Cell culture tested. Filtered at 0.1µm. DNase-, RNase-, and protease-free.
This product meets United States Pharmacopeia (USP) testing requirements, however it is not USP/EP certified.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100 °C (lit.)
Melting Pt: 0 (lit.)
Density: 1.000 g/ml at 3.98 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Water conductivity (packaged water) Passes test (result in µS/cm at 25°C)
Water conductivity (prior to packaging, stage 1) Passes test (result in µS/cm at temperature°C, this will vary by lot)
DNase None detected
Oxidizable substances Passes test (solution remains faintly pink)
Plating efficiency Passes test
Protease None detected
RNase None detected
Particulate matter in injections ≤3 Particles/ml ≥25 µm in size
Total organic carbon <500 ppb
Sterility Passes test
Bacterial endotoxin ≤0.025 EU/ml (per master product record)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR