Water, OmniSolv®, gradient grade, Supelco®

Supplier: MilliporeSigma

WX0004-NP20 WX0004-1 WX0004-6 WX0004P-1
CAWX0004-NP20EA 1068 CAD
CAWX0004-NP20 CAWX0004-1 CAWX0004-6 CAWX0004P-1
Water, OmniSolv®, gradient grade, Supelco®
Water

Meets reagent specifications for testing USP/NF monographs.

Formula: H₂O
Boiling Pt: 100 °C (lit.)
Melting Pt: 0 °C (lit.)
Density: 1.000 g/ml at 3.98 °C (lit.)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Molar Mass 18.02
Expiration Date 12 months from manufacture date
Color (APHA) 10 max
Filtered Through 0.2 µm Filter To pass test
Fluorescence (as Quinine Base) 100 ppt max
Gradient at 210 nm 0.002 AU max
Gradient at 254 nm 0.0005 AU max
Odor Odorless
Residue After Evaporation 1 ppm max
Specific Conductance 2 µmho/cm max
Total Purgeable Organics 5 ppb max
UV Abs. at 190 nm 0.010 AU max
UV Abs. at 200 nm 0.010 AU max
UV Abs. at 210 nm 0.010 AU max
UV Abs. at 220 nm 0.010 AU max
UV Abs. at 230 nm 0.010 AU max
UV Abs. at 240 nm 0.010 AU max
UV Abs. at 250 nm 0.005 AU max
UV Abs. at 280 nm 0.005 AU max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR