Water, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®

Supplier: MilliporeSigma

Synonyms: Hydrogen oxide

WX0001-1 WX0001-6
CAWX0001-1CS 2244 CAD
CAWX0001-1 CAWX0001-6
Water, OmniSolv® LC-MS LC-MS for LC-MS, Supelco®
Water

Clear liquid. For use in HPLC gradient analysis, GC, and spectrophotometry. Filtered through a 0.2µm element for purity.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now

Specification Test Results

Expiration date Exp 12 months from mfg date
Filtered through 0.2 µm filter To pass test
Fluorescence as quinine at 254 nm 1ppb max
Fluorescence as quinine at 365 nm 1ppb max
Gradient at 210 nm 0.002AU max
Gradient at 254 nm 0.0005AU max
Metals, suitable for LC/MS To pass test
MS purity, ESI(+), largest response as reserpine 50ppb max
MS purity, ESI(-), largest response, p-nitrophenol 50ppb max
Residue after evaporation 1ppm max
UV Abs. at 200 nm 0.01AU max
UV Abs. at 210 nm 0.01AU max
UV Abs. at 220 nm 0.01AU max
UV Abs. at 230 nm 0.01AU max
UV Abs. at 240 nm 0.01AU max
UV Abs. at 250 nm 0.005AU max
UV Abs. at 270 nm 0.005AU max
UV Abs. at 400-280 nm 0.005AU max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR