Water, OmniPur® for molecular biology nuclease-free, sterile, Calbiochem®

Supplier: MilliporeSigma

Synonyms: Hydrogen oxide

9801-100ML 9811-5L
CA9801EA 122.94 CAD
CA9801 CA9811 CA9812
Water, OmniPur® for molecular biology nuclease-free, sterile, Calbiochem®
Water

Intended for laboratory and manufacturing use only. Not for drug, food, or household use.


  • DNase, RNase and protease free
  • Sterile

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR