Water, OmniPur® Nuclease_free, sterile, DEPC_treated, Sigma-Aldrich®

Supplier: MilliporeSigma

Synonyms: Hydrogen oxide

9601-100ML 9602-500ML 9612-5L 9613-10L 9610-1L
CA9601EA 96.33 CAD
CA9601 CA9602 CA9612 CA9613 CA-EM9610L
Water, OmniPur® Nuclease_free, sterile, DEPC_treated, Sigma-Aldrich®
Water

Prepared with DEPC for RNA applications. DNase-, RNase-, and protease-free.

Formula: H₂O
MW: 18.02 g/mol
Boiling Pt: 100
Melting Pt: 0
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR