Vanadium(V) oxide ≥99.99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Vanadium pentoxide, di-Vanadium pentaoxide
10904-04
10904-09
10904-18
CAAA10904-04EA
98.43
CAD
CAAA10904-04
CAAA10904-09
CAAA10904-18
Vanadium(V) oxide ≥99.99%
Vanadium (V) oxide
Powder, 22 mesh. Purity based on metal contaminates only.
Purity: 99.99%
CAS No.: 1314-62-1
Molecular Formula: V2O5
Formula Weight: 181.88
Physical Form: -22 Mesh Powder
Density: 3.357
Appearance: Orange
Odor: Odorless
Melting Point: 690°
Boiling Point: 1750° dec.
Merck Reference: 14,9921
UN No.: 2862
MDL No.: MFCD00011457
Formula:
O₅V₂ MW: 181.88 g/mol Boiling Pt: 1750 (dec.) Melting Pt: 690 Density: 3.357 Storage Temperature: Ambient |
MDL Number:
MFCD00011457 CAS Number: 1314-62-1 EINECS: 215-239-8 UN: 2862 ADR: 6.1 III Merck Index: 14,9921 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...