Ethylene glycol ≥99% (by GC), BAKER ANALYZED® A.C.S. Reagent

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Ethane-1,2-diol, 1,2-Dihydroxy ethane, 1,2-Ethanediol, Glycol

CAJT9300-7 BAKR9300-R BAKR9300-03 9300-07 9300-33
CAJT9300-7EA 26475.05 CAD
CAJT9300-7 CAJT9300-R CAJT9300-1 CAJT9300-3 CAJT9300-33 JT9300-7 JT9300-33
Ethylene glycol ≥99% (by GC), BAKER ANALYZED® A.C.S. Reagent
Ethylene glycol

Achieve unsurpassed consistency, accurate results and industry-leading performance with solvents from Avantor Performance Materials. J.T.Baker® and Macron Fine Chemicals™ brands of solvents are available in sizes ranging from 500 mL to bulk packages containing hundreds of gallons. Our bulk solvent delivery capabilities are ideal for both pharmaceutical and biopharmaceutical production.

Formula: HOCH₂CH₂OH
MW: 62.07 g/mol
Boiling Pt: 196…198 °C (1013 hPa)
Melting Pt: -13
Density: 1.115 g/cm³ (20 °C)
Flash Pt: 116
Storage Temperature: Refrigerator
MDL Number: MFCD00002885
CAS Number: 107-21-1
EINECS: 203-473-3
REACH: 01-2119456816-28
Merck Index: 13,03832
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c4d1c97392851ba184f445f6ee912f84631f98d9&vid=2002300f174929a030d66d9f1394eb6bee89b7dc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Appearance Passes Test
Assay (HOCH₂CH₂OH) (by GC) ≥99 %
Assay (HOCH₂CH₂OH)(wet analysis) ≥99 %
Color (APHA) ≤10
Specific Gravity at 20°/20°C 1.114 - 1.116
Residue after Ignition ≤0.005 %
Acidity (as CH₃COOH) ≤0.01 %
Water (H₂O)(by Karl Fischer titrn) ≤0.2 %
Chloride (Cl) ≤5 ppm
Trace Impurities - Iron (Fe) ≤0.2 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR