Ethylenediamine ≥98.0%, BAKER ANALYZED®

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: 1,2-Ethanediamine, 1,2-Diaminoethane

Management of Change (MOC) category = N

BAKR9299-03 BAKR9299-01
CAJT9299-3CS 2793.77 CAD
CAJT9299-3 CAJT9299-1
Ethylenediamine ≥98.0%, BAKER ANALYZED®
Ethylenediamine

Lot analysis on label.

Formula: C₂H₈N₂
Boiling Pt: 117 °C (1013 hPa)
Melting Pt: 8.5 °C
Density: 0.898 g/cm³ (20 °C)
Flash Pt: 34 °C
Storage Temperature: Ambient
MDL Number: MFCD00008204
CAS Number: 107-15-3
EINECS: 203-468-6
UN: 1604
ADR: 8,II
Merck Index: 13,03829
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0cbd90391008b8df01337e52ed5760383c063baa&vid=9a49702fe7150a0d3cd56afb586972447ecb6f85&prid=web&clid=SAPDAM

Order Now

Specification Test Results

For Laboratory,Research,or Manufacturing Use
Assay (NH₂CH₂CH₂NH₂) ≥98.0 %
Density (g/mL) at 25°C 0.890 - 0.903
Heavy Metals (as Pb) ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR