Water, ultrapure HPLC
Supplier: Thermo Scientific Chemicals
Synonyms:
Hydrogen oxide
22934-K2
22934-K7
22934-M6
CAAA22934-K2EA
86.13
CAD
CAAA22934-K2
CAAA22934-K7
CAAA22934-M6
Water, ultrapure HPLC
Water
CAS No.: 7732-18-5
Molecular Formula: H2O
Formula Weight: 18.02
Physical Form: Liquid
Density: 1.000
Refractive Index: 1.3330
Odor: Odorless
Melting Point: 0°
Boiling Point: 100°
Merck Reference: 14,10039
MDL No.: MFCD00011332
Formula:
H₂O MW: 18.02 g/mol Boiling Pt: 100 Melting Pt: 0 Density: 1.000 Storage Temperature: Ambient |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 EINECS: 231-791-2 Merck Index: 14,10039 |
Specification Test Results
Appearance (Color) | Clear colorless |
Form | Liquid |
Assay (unspecified) | Specific Conductance at 25°C and time packaging: 1.5 microS/cm, max |
Optical Absorbance at | 400 nm 0.005 AU, max- at 300 nm 0.005 AU, max- at 250 nm 0.005 AU, max- at 200 nm 0.010 AU, max- at 190 nm 0.010 AU, max |
Residue on Evaporation | 2 ppm, Max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...