Water, NERL™, Reagent Grade

Supplier: Thermo Fisher Scientific

Synonyms: Hydrogen oxide

9800-1 9800-3 9800-3
48218-708CS 532.74 CAD
48218-708 72060-096 CA72060-096L
Water, NERL™, Reagent Grade
Water

Meets CAP and NCCLS specifications for reagent grade, Type 1 water at time of preparation. Ideal for reconstituting chemistry or coagulation products, preparing analytical standards, or rinsing delicate electrodes. Prepared at 18 megohm/cm specific resistance using reverse osmosis, mixed bed deionization, activated carbon filtration, and final filtration to 0.2µm. Contains no preservatives and is essentially free of soluble organics and inorganic particulate contaminants. Certificate of Analysis includes over 60 parameters and analytes.

Formula: H₂O
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: 0 °C
Density: 1 g/cm³ (4 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011332
CAS Number: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR