Water, ARISTAR® PLUS for HPLC low TOC, VWR Chemicals BDH®
Supplier: VWR International
Synonyms:
Hydrogen oxide
10901-VWNA63
10901-VWNA43
10901-VWNA63
87003-652CS
2151.07
CAD
87003-650
87003-652
CA87003-650
Water, ARISTAR® PLUS for HPLC low TOC, VWR Chemicals BDH®
Water
For LC, HPLC, and trace organic analysis.
- Used in analytical and preparative separations
- Suitable for gradient and isocratic conditions
- Filtered through a 0.5 μm filter to remove particulates
LC Gradient Suitability Tested:
Column: 25 cm C18
Flow rate: 2 ml/min
Conditions: 40 ml of water are passed through the column and eluted with a water acetonitrile gradient at 5% per min.
Maximum Absorbance: 0.002 at 254 nm
Formula:
H₂O MW: 18.02 g/mol Boiling Pt: 100 °C (1013 hPa) Melting Pt: 0 °C Density: 1 g/cm³ (4 °C) |
MDL Number:
MFCD00011332 CAS Number: 7732-18-5 Merck Index: 12,10175 |
Specification Test Results
Color | max. 5 APHA |
Residue after Evaporation | max. 1 ppm |
Fluorescence (as Quinine) | max. 100 ppt |
Resistivity (at time of bottling) | min. 18 MΩ |
Total Organic Carbon | max. 20 ppb |
LC Gradient Suitability Test: | to pass test |
Optical Absorbance at 200 nm | max. 0.01 |
Optical Absorbance at 254 nm | max. 0.005 |
Optical Absorbance at 300 nm | max. 0.005 |
Optical Absorbance at 400 nm | max. 0.005 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...