EDTA disodium salt concentrate for dilution to 0.1 M in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Synonyms:
Edetic acid disodium salt
, Disodium dihydrogen ethylenediaminetetraacetate
, Ethylenediaminetetra-acetic acid disodium salt
, Idranal III
, Kestranal® 2S
, Ethylenedinitrilotetra-acetic acid disodium salt
Management of Change (MOC) category = N
J.T. Baker®
4653-01
BAKR4653-01
BAKR4653-01
CAJT4653-1CS
1258.82
CAD
CAJT4653-1
JT4653-1-U
JT4653-1
EDTA disodium salt concentrate for dilution to 0.1 M in aqueous solution, DILUT-IT®, Laboratory Reagent, J.T.Baker®
EDTA disodium salt
(1/10 mole=33.62 g C₁₀H₁₄N₂Na₂O₈)
Formula:
C₁₀H₁₄N₂Na₂O₈ MW: 336.21 g/mol |
MDL Number:
MFCD00070672 CAS Number: 139-33-3 Merck Index: 12,03556 |
Specification Test Results
For laboratory, research or manufacturing use. | |
Molarity (by titrimetry) | Passes Test |
(Makes 0.1M solution after dilution to 1000 ml) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...