KIMAX® Filtering Flasks with Capacity Scale, Without Tubulation, Kimble Chase
Supplier: DWK Life Sciences (KIMBLE)
27050-1000 27050-2000 27050-4000 27050-500
89001-790CS 181.28 CAD
89001-790 89001-792 89001-794 89001-796
KIMAX® Filtering Flasks with Capacity Scale, Without Tubulation, Kimble Chase
Flasks Erlenmeyer Flasks
Designed for maximum strength.
  • Necks are tooled for a uniform stopper fit with a thick flat flange
  • Durable white ceramic enamel scale indicates approximate volume
  • Without side tubulation
The 500 ml flask is designed from ASTM Specification E1406, Type I requirements.