Ethylene glycol ≥99%, Supelco®

Supplier: MilliporeSigma

Synonyms: 1,2-Ethanediol1,2-Dihydroxy ethaneEthane-1,2-diolGlycol

EX0564-1 EX0564-1
CAEX0564-1CS 6642.88 CAD
CAEX0564-1 EM-EX0564-1
Ethylene glycol ≥99%, Supelco®
Ethylene glycol
Formula: HOCH₂CH₂OH
MW: 62.07 g/mol
Boiling Pt: 196…198 °C (1013 hPa)
Melting Pt: –13 °C
Density: 1.115 g/cm³ (20 °C)
Flash Pt: 116 °C
MDL Number: MFCD00002885
CAS Number: 107-21-1
Merck Index: 13,03832
https://ca.vwr.com/stibo/web/std.lang.all/43/75/734375.jpg

Order Now

Specification Test Results

Assay (by Mole/Mole) 99% min.
Water (H2O) 0.2% max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR