Ethanol denatured 92-94% for histology, for biochemistry, Supelco®

Supplier: MilliporeSigma
Danger

EX0285-1 EX0285-3
CA-EX0285-1EA 1702.18 CAD
CA-EX0285-1 CA-EX0285-3
Ethanol denatured 92-94% for histology, for biochemistry, Supelco®
Ethanol denatured

100 volumes SD Alcohol #1 (1 volume Ethyl Acetate, 1 volume Hydrocarbon solvent, 1 volume MIBK)


  • Total alcohol: 95-99%
  • Ethyl alcohol: 92-94%

Formula: H₃CCH₂OH
Flash Pt: 13
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Assay (Ethanol) 92-94%
Assay (Ethyl acetate) 1-2%
Assay (Methanol) 3-5%
Acidity (as acetic acid) 0.005% max
Color (APHA) 20 max
Form Clear liquid
Infrared spectrum Conforms to standard
Nonvolatile residue 0.0025% max
Specific Gravity at 20/20°C 0.789-0.793
Water (H2O) 0.5% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR