Ethanol denatured 95% for histology, for biochemistry (denatured with methanol and ethyl acetate), Supelco®

Supplier: MilliporeSigma

Synonyms: Ethyl alcohol denatured

EX0280-1 EX0280-30 EX0280-30 EX0280-3 EX0280-30
CAEM-EX0280-30EA 3309.36 CAD
CAEM-EX0280-30 EM-EX0280-3 TXEX028030MIC EM-EX0280-1 EM-EX0280-30
Ethanol denatured 95% for histology, for biochemistry (denatured with methanol and ethyl acetate), Supelco®
Ethanol denatured

100 volumes SD Alcohol #1 (100 volumes Ethanol, 5 volumes Methanol)


  • Total alcohol: 89-94%
  • Ethyl alcohol: 86-89%
  • Denaturant: 1-2% ethyl acetate and 3-5% methyl alcohol

Formula: H₃CCH₂OH
MW: 46.07 g/mol
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Assay (Ethanol) 86.0-89.0%
Assay (Ethyl acetate) 1-2%
Assay (Methanol) 3-5%
Assay (Water) 6-8%
Acidity (as acetic acid) 0.005% max
Color (APHA) 10 max
Form Clear liquid
Infrared spectrum Conforms to standard
Nonvolatile residue 0.0025% max
Specific Gravity at 20/20°C 0.809-0.813

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR