Nalgene® Erlenmeyer Flasks, Polycarbonate, Thermo Scientific
Supplier: Thermo Fisher Scientific
4103-0125 4103-0250
29136-933CS 633.7 CAD
29136-933 29136-944
Nalgene® Erlenmeyer Flasks, Polycarbonate, Thermo Scientific
Flasks Erlenmeyer Flasks
Graduated flasks feature excellent clarity, toughness, and strength.
Molded-in graduations. Autoclavable, but repeated autoclaving may reduce mechanical strength.