Ethyl acetate 99,8 HPLC, Supelco®

Supplier: MilliporeSigma
Danger

Synonyms: Acetic acid ethyl ester

EX0245-6 EX0245P-1 EX0245-1
CAEX0245-6EA 267 CAD
CAEX0245-6 CAEX0245P-1 CAEX0245-1
Ethyl acetate 99,8 HPLC, Supelco®
Ethyl acetate

HPLC, Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: CH₃COOC₂H₅
MW: 88.11 g/mol
Boiling Pt: 77.1
Melting Pt: -83
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3 II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.8% min
Appearance Clear liquid, free from particulates
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Form Liquid
Infrared spectrum Conforms to standard
Residue after evaporation 5ppm max
Substances darkened by sulfuric acid To pass test
Titrable acid 0.0009meq/g max
UV Abs. at 255 nm 1.00AU max
UV Abs. at 257 nm 0.50AU max
UV Abs. at 263 nm 0.10AU max
UV Abs. at 275 nm 0.05AU max
UV Abs. at 300 - 400 0.01AU max
UV Cut-off 255nm max
Water (H2O) 0.05% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR