Ethyl acetate ≥99.9%, OmniSolv® for biosynthesis, Supelco®

Supplier: MilliporeSigma
Danger

EX0242-5 EX0242-1
CAEX0242-5EA 87 CAD
CAEX0242-5 CAEX0242-1
Ethyl acetate ≥99.9%, OmniSolv® for biosynthesis, Supelco®
Ethyl acetate

Biosynthesis

Formula: CH₃COOC₂H₅
Boiling Pt: 77.1 °C (1013 hPa)
Melting Pt: –83 °C
Density: 0.902 g/cm³ (20 °C)
Flash Pt: –4 °C
Storage Temperature: Ambient
MDL Number: MFCD00009171
CAS Number: 141-78-6
EINECS: 205-500-4
UN: 1173
ADR: 3,II
REACH: 01-2119475103-46
Merck Index: 13,03792
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=226aecffe42ffab125597552eff0ef7f88ef9983&vid=f5707ce0faee09ca10700419252c5841e0fbc01a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay (GC) 99.9% min
Color (APHA) 10 max
Filtered through 0.2 µm filter To pass test
Fluorescence (as quinine base) 250ppt max
Form Clear liquid
Infrared spectrum Conforms to standard
Residue after evaporation 1ppm max
Titrable acid 0.8µeq/g max
UV Abs. at 255 nm 1.00AU max
UV Abs. at 257 nm 0.35AU max
UV Abs. at 263 nm 0.06AU max
UV Abs. at 275 nm 0.02AU max
UV Abs. at 300 nm 0.01AU max
UV Cut-off 255nm max
Water (H2O) 100ppm max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR