Diethyl ether, anhydrous ≥99.0% (by GC) stabilized, GR ACS, Supelco®
Supplier: MilliporeSigma
|
Danger
|
EX0190-20
EX0190-5
EX0190-3
EX0190-4
EX0190-6
EX0190-8
EX0190-9
CAEX0190-20EA
4847
CAD
CAEX0190-20
CAEX0190-5
CAEX0190-3
CAEX0190-4
CAEX0190-6
CAEX0190-8
CAEX0190-9
Diethyl ether, anhydrous ≥99.0% (by GC) stabilized, GR ACS, Supelco®
Diethyl ether
Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs
Formula:
(CH₃CH₂)₂O Boiling Pt: 34.6 °C (lit.) Melting Pt: –116 °C (lit.) Density: 0.706 g/ml (25°C) (lit.) Flash Pt: –40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011646 CAS Number: 60-29-7 EINECS: 200-467-2 UN: 1155 ADR: 3,I REACH: 01-2119535785-29 Merck Index: 12,03852 |
|
Specification Test Results
Expiration date | Exp 24 months from mfg date |
Assay (GC) | 99.0% min |
Alcohol(C2H5OH) (limit about 0.05%) | To pass test |
Carbonyl (as HCHO) | 0.001% max |
Color (APHA) | 10 max |
Form | Clear liquid |
Peroxide (as H2O2) | 1.0ppm max |
Residue after evaporation | 0.001% max |
Titrable acid | 0.0002meq/g max |
Water (H2O) | 0.03% max |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...