Diethyl ether ≥98% stabilized, GR ACS, Supelco®

Supplier: MilliporeSigma
Danger

EX0185-3 EX0185-4 EX0185-5 EX0185-6 EX0185-9 EX0185-8
CAEX0185-3CS 2717 CAD
CAEX0185-3 CAEX0185-4 CAEX0185-5 CAEX0185-6 CAEX0185-9 CAEX0185-8
Diethyl ether ≥98% stabilized, GR ACS, Supelco®
Diethyl ether

Meets ACS Specifications, Meets Reagent Specifications for testing USP/NF monographs

Formula: (CH₃CH₂)₂O
Boiling Pt: 35 °C (1013 hPa)
Melting Pt: –123 °C
Density: 0.7135 g/cm³ (20 °C)
Flash Pt: –40 °C
Storage Temperature: Ambient
MDL Number: MFCD00011646
CAS Number: 60-29-7
EINECS: 200-467-2
UN: 1155
ADR: 3,I
REACH: 01-2119535785-29
Merck Index: 12,03852
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16f2debd1f18f9bff9952c4a6e5d599a67f94e61&vid=3dd6c545e58e36e1004e828026523fb0e3bcbbcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Expiration date Exp 24 months from mfg date
Assay (Et2O + EtOH) 98.0% min
Alcohol (C2H5OH) About 2
Carbonyl (as HCHO) 0.001% max
Color (APHA) 10 max
Density at 20°C 0.714-0.718
Form Clear liquid
Peroxide (as H2O2)* 1ppm max
Residue after evaporation 0.001% max
Titrable acid 0.0002meq/g max
Water (H2O) 0.5% max


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR