Diethyl ether ≥99.9%, OmniSolv®, Supelco®

Supplier: MilliporeSigma

Synonyms: Ether2,2'-OxybisethaneEthoxyethaneEthyl Ether2,2'-Oxydiethane

EX0182-1 EX0182-1
CAEX0182-1CS 2329.63 CAD
CAEX0182-1 EM-EX0182-1
Diethyl ether ≥99.9%, OmniSolv®, Supelco®
Diethyl ether

Clear liquid. Suitable for residue analysis, HPLC, and spectrophotometry.

Formula: (CH₃CH₂)₂O
MW: 74.12 g/mol
Boiling Pt: 35 °C (1013 hPa)
Melting Pt: –123 °C
Density: 0.7135 g/cm³ (20 °C)
Flash Pt: –40 °C
MDL Number: MFCD00011646
CAS Number: 60-29-7
UN: 1155
ADR: 3,I
Merck Index: 12,03852
https://ca.vwr.com/stibo/web/std.lang.all/02/58/730258.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR