Nalgene® Square Media Bottles with Septum Closure, PETG, Sterile, Thermo Scientific
Supplier: Thermo Fisher Scientific
342023-0060 342023-0125 342023-0500 342023-1000
46600-586CS 1178.11 CAD
46600-586 46600-588 46600-590 46600-592
Nalgene® Square Media Bottles with Septum Closure, PETG, Sterile, Thermo Scientific
Bottles Cell Culture Bottles
Ideal for packaging of biological and diagnostic reagents that require aseptic dispensing or addition of supplements by syringe.
  • Noncytotoxic
  • Leakproof
  • Sterile
  • Offered in variety of sizes
The silicone/PTFE closure with septum provides a PTFE fluid contact surface that is inert and nonstick to ensure fluid integrity. Bottles and closures are sterile to 10 to 6 sterility assurance level.
Certifications: Biologically tested bottle and septum closure meet USP Class VI and European Pharmacopeia Abnormal Toxicity standards.
Ordering information: More Product Information