EDTA disodium salt dihydrate solution 0.5 M, OmniPur®, sterile, Millipore®

Supplier: MilliporeSigma

4056-500ML 4055-100ML
CA4056EA 1010 CAD
CA4056 CAEM4055
EDTA disodium salt dihydrate solution 0.5 M, OmniPur®, sterile, Millipore®
EDTA disodium salt dihydrate

For molecular biology. Prepared with diethyl pyrocarbonate for RNA applications. DNase-, RNase-, and protease-free.

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number: MFCD00150037
CAS Number: 6381-92-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2f89002087c5d4d84af5dded62c87287b1774fab&vid=0ef4c3febbcc8a20569606067799db31f2540748&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR