EDTA disodium salt dihydrate ≥99.0%, OmniPur® for biotechnology, Millipore®

Supplier: MilliporeSigma

Synonyms: Ethylenediaminetetraacetic acid disodium dihydrateEthylenediaminetetra-acetic acid disodium salt dihydrateEdetate disodium dihydrateEthylenediaminetetraacetic acid, disodium salt, dihydrateEthylenediaminetetraacetic acid disodium salt dihydrateKestranal® 2S 2 H2OEthylenedinitrilotetra-acetic acid disodium salt dihydrateTitriplex IIIKestranal® 2S dihydrateEdetic acid disodium salt dihydrateDisodium dihydrogen ethylenediaminetetraacetate dihydrateSequestrene Na₂

4050-1KG 4010-250GM 4050-1KG
CA-EM4050EA 908.69 CAD
EM-4010 EM-4050 CA-EM4050
EDTA disodium salt dihydrate ≥99.0%, OmniPur® for biotechnology, Millipore®
EDTA disodium salt dihydrate

For molecular biology. DNase-, RNase-, protease-, and phosphatase-free.

Formula: C₁₀H₁₄N₂Na₂O₈·2H₂O
MW: 372.24 g/mol
Melting Pt: 255 °C
MDL Number: MFCD00150037
CAS Number: 6381-92-6
Merck Index: 13,03543
https://ca.vwr.com/stibo/web/std.lang.all/99/56/2299956.jpg

Order Now

Specification Test Results

Max. UV Absorbance at 260 nm 0.2
Max. UV Absorbance at 280 nm 0.02%
Assay 99.0% min.
Heavy Metals (Pb) 0.0005% max.
Loss on Drying 9.0–10.0% max.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR