Microwave/Refrigerator-Freezers Combinations with Allocator
Supplier: FELIX STORCH, INC./SUMMIT
MRF34BSSA MRF66BKA MRF73PLA
77442-218EA 2158.6 CAD
77442-218 77442-220 77442-224
Microwave/Refrigerator-Freezers Combinations with Allocator
Refrigerators
Microwave/Refrigerator-Freezer combination with allocator with one-plug operation, ideal for hotels and other settings looking to maximize user convenience. The allocator will temporarily disable the power of the refrigerator while the microwave is in use, resuming after the cooking cycle is complete in order to protect against power surges.
  • USB ports
  • Cycle defrost
  • Reversible doors (refrigerator)
  • Sealed back
  • Stainless steel handle (microwave)
  • Mute function
  • Eco mode
  • Thin-line design
Environmentally friendly hydrocarbon-based refrigerant for a greener footprint that meets current CARB regulations.
Certifications: CARB compliant.
Packaging: Ships on one pallet in three boxes.