Ethyl acetate 99,5, Purified Plus® ACS organic_synthesis, LC_Preparative, Burdick & Jackson™
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
Synonyms:
Acetic acid ethyl ester
PP100-19
PP100-204
CABJPP100-19EA
952.98
CAD
CABJPP100-19
CABJPP100-204
Ethyl acetate 99,5, Purified Plus® ACS organic_synthesis, LC_Preparative, Burdick & Jackson™
Ethyl acetate
For specialty organic synthesis, preparative scale liquid chromatography, glassware cleaning, and applications requiring ACS reagent-grade solvent. The 54gal. drum is individually analyzed with certificate on label.
- Low residue
- Purified solvent
- ACS General Use certified
Formula:
CH₃COOC₂H₅ MW: 88.11 g/mol Boiling Pt: 77.1 Melting Pt: -83 Density: 0.9006 g/cm³ (20 °C) Flash Pt: –4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009171 CAS Number: 141-78-6 EINECS: 205-500-4 UN: 1173 ADR: 3 II REACH: 01-2119475103-46 Merck Index: 13,03792 |
|
Specification Test Results
Assay by GC Analysis | 99.9 % min. |
Color (APHA) | 10 max. |
Refractive Index (20ºC) | 1.3713 - 1.3733 |
Residue after Evaporation | 5 mg/L max. |
Substances Darkened by Sulfuric Acid | Passes test |
Titrable Acid | 0.9 µeq/g max. |
Water by Karl Fischer Titration | 0.1 % max. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...