Diethyl ether ≥98.5% stabilized ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
Supplier: HONEYWELL BURDICK & JACKSON CA
|
Danger
|
Synonyms:
Ether
, 2,2'-Oxybisethane
, Ethoxyethane
, Ethyl Ether
, 2,2'-Oxydiethane
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
LP108-4
LP108-19
LP108-4
LP108-19
CABJLP108D19EA
1896.74
CAD
BJLP108D19
CABJLP108D19
CABJLP108-4
BJLP108-4
Diethyl ether ≥98.5% stabilized ACS for organic synthesis, for preparative liquid chromatography, general laboratory use
Diethyl ether
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Formula:
(CH₃CH₂)₂O MW: 74.12 g/mol Boiling Pt: 35 °C Melting Pt: –117.4 °C Density: 0.713 g/cm³ (20 °C) Flash Pt: –45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011646 CAS Number: 60-29-7 UN: 1155 ADR: 3,I Merck Index: 12,03852 |
Specification Test Results
Alcohol (as CH3CH2OH) | Passes test |
Appearance | Passes test |
Assay by GC Analysis | 98 % min. |
Carbonyl (as HCHO) | 0.001 % max. |
Color (APHA) | 10 max. |
Peroxides (as H₂O₂) at time of packaging | 1 ppm max. |
Residue after Evaporation | 0.001 % max. |
Titrable Acid | 0.2 µeq/g max. |
Water by Karl Fischer Titration | 0.03 % max. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...